Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIX3/SIX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575220
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The SIX proteins (sine oculis) are a family of homeodomain transcription factors that share a conserved DNA binding domain. Two of these family members Six3 and Six6 (also designated Optx2 and Six9) are required for the specification and proliferation of the eye field in vertebrates, and, therefore, are the vertebrate homologues most closely related to the Drosophila sine oculis protein, which has an essential role in controlling compound eye development. Six3 and Six6 expression largely overlap during development of specific tissues, such as retina, hypothalamus, and pituitary. Deletion of Six3 may be associated with HPE2 disorder, a common, severe malformation of the brain that results from incomplete cleavage of the forebrain during early embryogenesis.
Specifications
SIX3/SIX6 | |
Polyclonal | |
Unconjugated | |
SIX3 | |
E130112M24Rik; homeobox protein SIX3; Homeobox protein SIX6; homeodomain protein OPTX2; HPE2; MCOPCT2; ODRMD; optic homeobox 2; OPTX2; Sine oculis homeobox homolog 3; sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 3; sine oculis homeobox-like protein 6; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; sine oculis-related homeobox 6; sine oculis-related homeobox 6 homolog; SIX homeobox 3; SIX homeobox 6; SIX3; Six3a; Six3alpha; Six3b; Six3beta; Six6; Six9; Six9 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20473, 20476, 314221, 4990, 6496, 78974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O95343, O95475, Q62233, Q9QZ28 | |
SIX3, SIX6 | |
Synthetic peptide corresponding to amino acids 201-250 of Human Six3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction