Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIX4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SIX4 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SIX4 Polyclonal specifically detects SIX4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SIX4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
51804 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DGSTFTSESTTVQQGKVFLSSLAPSAVVYTVPNTGQTIGSVKQEGLERSLVFSQLMPVNQNAQVNANLSSENISGSGLHPLASSLVNVSPTH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Rabbit | |
Human | |
AREC3, homeobox protein SIX4, MGC119450, MGC119452, MGC119453, sine oculis homeobox (Drosophila) homolog 4, Sine oculis homeobox homolog 4, sine oculis homeobox homolog 4 (Drosophila), SIX homeobox 4 | |
SIX4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title