Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556861 Supplier Invitrogen™ Supplier No. PA556861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGSTFTSEST TVQQGKVFLS SLAPSAVVYT VPNTGQTIGS VKQEGLERSL VFSQLMPVNQ NAQVNANLSS ENISGSGLHP LASSLVNVSP TH Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.

SIX4 gene ontology annotations related to this gene include anatomical structure development; embryonic cranial skeleton morphogenesis; fungiform papilla morphogenesis; generation of neurons; inner ear morphogenesis; male gonad development; male sex determination; metanephric mesenchyme development; myoblast migration; myotome development; negative regulation of neuron apoptotic process; negative regulation of satellite cell differentiation; negative regulation of transcription, DNA-templated; olfactory placode formation; pharyngeal system development; positive regulation of branching involved in ureteric bud morphogenesis; positive regulation of ureteric bud formation; protein localization to nucleus; regulation of branch elongation involved in ureteric bud branching; regulation of epithelial cell proliferation; regulation of protein localization; regulation of synaptic growth at neuromuscular junction; sarcomere organization; skeletal muscle fiber differentiation; thymus development; trigeminal ganglion development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX4
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIX4
Gene Accession No. Q9UIU6
Gene Alias AI047561; Arec3; homeobox protein SIX4; Sine oculis homeobox homolog 4; sine oculis-related homeobox 4; sine oculis-related homeobox 4 homolog; SIX homeobox 4; Six4; Skeletal muscle-specific ARE-binding protein AREC3; TrexBF
Gene Symbols SIX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIX4. Recombinant protein control fragment (Product #RP-94975).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.