Learn More
SIX4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303220A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
The function of this protein remains unknown.Specifications
SIX4 | |
Polyclonal | |
Unconjugated | |
SIX4 | |
AREC3, sine oculis homeobox (Drosophila) homolog 4, sine oculis homeobox homolog 4, sine oculis homeobox homolog 4 (Drosophila) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51804 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UIU6 | |
SIX4 | |
Between 731 and 781 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.