Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590859 Supplier Invitrogen™ Supplier No. PA590859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATLPAEPSA GPAAGGEAVA AAAATEEEEE EARQLLQTLQ AAEGEAAAAA GAGAGAAAAG AEGPGSPGVP GSPPEAASEP PTGLRFSPEQ VACVCEALLQ AGHAGRLSRF LGALPPAERL RGSDPVLRAR ALVAFQRGEY AELYRLLESR PFPAAHHAFL QDLYLRARYH; Positive Samples: Mouse eye, Rat eye; Cellular Location: Cytoplasm, Nucleus.

Six5 (homeobox protein SIX5), also known as SIX5, BOR2 or DMAHP (DM locus-associated homeodomain protein), is a transcription factor that is expressed in various structures of the adult eye. Localized to the cytoplasm in early development and to the nucleus in the later stages of development, Six5 is involved in regulation of organogenesis and in maintenance of retinal formation. Six5 is able to bind the 5'-TCA[AG][AG]TTNC-3' DNA sequence found in the myogenin and IGFBP5 promoters and, through this binding, can control transcription of the associated mRNA. Six5 is regulated via association with DACH1 (dachshund homolog 1) and is co-activated by the EYA (eyes absent) proteins. Defects in the gene encoding Six5 are the cause of branchiooto- renal syndrome type 2 (BOR2), an autosomal disorder characterized by hearing loss, a deep overbite and myopia. Two isoforms exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Six5
Gene Accession No. P70178
Gene Alias BOR2; DM locus-associated homeodomain protein; DM locus-associated homeodomain protein homolog; DMAHP; dystrophia myotonica-associated homeodomain protein; homeobox protein SIX5; MDMAHP; Sine oculis homeobox homolog 5; sine oculis-related homeobox 5; sine oculis-related homeobox 5 homolog; SIX homeobox 5; Six5; TrexBF
Gene Symbols Six5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human SIX5 (NP_7870712).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20475, 308406
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.