Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540534 Supplier Invitrogen™ Supplier No. PA540534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PAACEALNKN ESVLRARAIV AFHGGNYREL YHILENHKFT KESHAKLQAL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in eye development. Defects in this gene are a cause of isolated microphthalmia with cataract type 2 (MCOPCT2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIX6
Gene Accession No. O95475
Gene Alias Homeobox protein SIX6; homeodomain protein OPTX2; MCOPCT2; ODRMD; optic homeobox 2; Optx2; sine oculis homeobox 6; sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 6; sine oculis-related homeobox 6; sine oculis-related homeobox 6 homolog; SIX homeobox 6; Six6; SIX9; Six9 protein
Gene Symbols SIX6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SIX6 (aa 41-90).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4990
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.