Learn More
Invitrogen™ SIX6 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SIX6 protein.
Specifications
Specifications
| Antigen | SIX6 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SIX6 |
| Gene Accession No. | O95475, Q9QZ28 |
| Gene Alias | Homeobox protein SIX6; homeodomain protein OPTX2; MCOPCT2; ODRMD; optic homeobox 2; Optx2; sine oculis homeobox 6; sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 6; sine oculis-related homeobox 6; sine oculis-related homeobox 6 homolog; SIX homeobox 6; Six6; SIX9; Six9 protein |
| Gene Symbols | SIX6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.