Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551573
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA551573 100 μL
1 options

Catalog No. PIPA551573

Supplier: Invitrogen™ PA551573

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YQEAEKLRGR PLGPVDKYRV RKKFPLPRTI WDGEQKTHCF KERTRNLLRE WYLQDPYPNP SKKRELAQAT GLTPTQVGNW FKNRRQRDRA AAAKNRLQQQ VLSQGSGRAL RAEGDGTPEV LGVATSPAAS LSSKAATSAI SITSSDSECD I Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in eye development. Defects in this gene are a cause of isolated microphthalmia with cataract type 2 (MCOPCT2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIX6
Gene Accession No. O95475
Gene Alias Homeobox protein SIX6; homeodomain protein OPTX2; MCOPCT2; ODRMD; optic homeobox 2; Optx2; sine oculis homeobox 6; sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 6; sine oculis-related homeobox 6; sine oculis-related homeobox 6 homolog; SIX homeobox 6; Six6; SIX9; Six9 protein
Gene Symbols SIX6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIX6. Recombinant protein control fragment (Product #RP-88903).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.