Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SK 216
GREENER_CHOICE

Catalog No. 618750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-875-0 50 mg
61-871-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-875-0 Supplier Tocris Bioscience™ Supplier No. 6187/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Plasminogen activator inhibitor-1 (PAI-1) inhibitor

SK 216 is a plasminogen activator inhibitor-1 (PAI-1) inhibitor. Inhibits VEGF-induced migration and tube formation in HUVECs. Also attenuates TGF-β dependent epithelial-mesenchymal transition and fibroblast to myofibroblast differentiation in vitro. Reduces size and weight of subcutaneous tumors, and inhibits metastasis and angiogenesis in vivo. Reduces bleomycin-induced pulmonary fibrosis in vivo. Orally bioavailable.

Specifications

Chemical Name or Material 2-[5-[[6-[5-(1,1-Dimethylethyl)-2-benzoxazolyl]-2-naphthalenyl]oxy]pentyl]propanedioic acid disodium salt
CAS 654080-03-2
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C29H29NO6Na2
Formula Weight Observed MW: 533.52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.