Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
SK 575 is a potent PARP1 Degrader (PROTAC®; DC50 ≤ 6.72 nM, >99% PARP1 degradation at 10 nM). It comprises the PARP1/2 inhibitor Olaparib (Cat. No. 7579), joined by a linker to cereblon/cullin 4A ligand Thalidomide (Cat. No. 0652). SK 575 inhibits the growth of various cancer cell lines bearing BRCA1/2 mutations with IC50 values in the nanomolar range. It effectively reduces PARP1 protein levels in mouse SW620 tumor xenograft models. SK 575 dose-dependently potentiates the antitumor activity of Temozolomide (Cat. No. 2706) and Cisplatin (Cat. No. 2251) in vivo.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethyl)-12-(4-(2-fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzoyl)piperazin-1-yl)-12-oxododecanamide |
| CAS | 2523016-96-6 |
| Quantity | 5 mg |
| Target | PARP |
| Molecular Formula | C47H53FN8O8 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

