Learn More
Invitrogen™ SKA1 Polyclonal Antibody

Description
A suggested positive control is A549 cell lysate. PA5-20817 can be used with blocking peptide PEP-0931.
Specifications
Specifications
| Antigen | SKA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SKA1 |
| Gene Accession No. | B0BN28, Q96BD8, Q9CPV1 |
| Gene Alias | 2810433K01Rik; AV117428; C18orf24; RGD1310784; SKA1; spindle and kinetochore associated complex subunit 1; spindle and kinetochore-associated protein 1; spindle and KT (kinetochore) associated 1 |
| Gene Symbols | SKA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.