Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563689 Supplier Invitrogen™ Supplier No. PA563689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAEVDKLELM FQKAESDLDY IQYRLEYEIK TNHPDSASEK NPVTLLKELS VIKS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.

Upon entry into mitosis, the cell's microtubule (MT) network forms the mitotic spindle, allowing the segregation of paired chromosomes. Proteinaceous structures on centromeric chromatin termed kinetochores (KT) are essential for the proper attachment of the chromosomes to the spindle MTs. A recently discovered spindle and kinetochore complex, comprised of proteins SKA1, SKA2, and SKA3, has been found to be required for stable KT-MT interactions and timely anaphase onset. Depletion of either SKA1 or SKA2 by siRNA results in the loss of both proteins from the KT, but does not impact overall KT structure. Cells depleted of the SKA complex undergo a prolonged checkpoint-dependent delay in a metaphase-like state, indicating the importance of the SKA complex in the maintenance of the metaphase plate and spindle checkpoint silencing. SKA2 has also been shown to interact with glucocorticoid receptors and to be involved in glucocorticoid signaling and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKA2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SKA2
Gene Accession No. Q8WVK7
Gene Alias 1110001A07Rik; C78640; FAM33A; family with sequence similarity 33, member A; Protein FAM33A; RGD1307084; Ska2; spindle and kinetochore associated complex subunit 2; spindle and kinetochore-associated protein 2; spindle and KT (kinetochore) associated 2
Gene Symbols SKA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SKA2. Recombinant protein control fragment (Product #RP-100886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348235
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.