Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKA3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8066 Supplier Bioss Supplier No. BS7848R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Upon entry into mitosis, the cell′s microtubule (MT) network forms the mitotic spindle, allowing the segregation of paired chromosomes. Proteinaceous structures on centromeric chromatin termed kinetochores (KT) are essential for the proper attachment of the chromosomes to the spindle MTs. A recently discovered spindle and kinetochore complex, comprised of proteins SKA1, SKA2, and SKA3, has been found to be required for stable KT-MT interactions and timely anaphase onset. Like with SKA1 or SKA2, depletion of SKA3 by siRNA delays anaphase transition, resulting in a prolonged a metaphase-like state. These SKA3-depleted cells accumulate high levels of the checkpoint protein Bub1 at kinetochores, suggesting the SKA complex plays a key role in spindle checkpoint silencing and the maintenance of chromosome cohesion in mitosis.

Specifications

Antigen SKA3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SKA3
Gene Accession No. Q8IX90
Gene Alias C13orf3; F630043A04Rik; RAMA1; RGD1307201; RP11-101P17.12-002; Ska3; spindle and kinetochore associated complex subunit 3; Spindle and kinetochore-associated protein 3
Gene Symbols SKA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SKA3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221150
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.