Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKAP Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306036MFV610
Description
SKAP Polyclonal antibody specifically detects SKAP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SKAP | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
C15orf23, chromosome 15 open reading frame 23, FLJ14502, HSD11, MGC141728, MGC141729, putative TRAF4-associated factor 1, SKAP, small kinetochore associated protein, TRAF4 associated factor 1, TRAF4AF1 | |
Produced in rabbits immunized with E. coli-derived Human SKAP fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
90417 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction