Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$670.00
Specifications
Antigen | SKAP |
---|---|
Dilution | Simple Western 1:20, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SKAP Polyclonal specifically detects SKAP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SKAP | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
90417 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TVYSLQPPSALSGGQPADTQTRATSKSLLPVRSKEVDVSKQLHSGGPENDVTKITKLRRENGQMKATDTATRRNVRKGYKPLSKQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Simple Western 1:20, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
C15orf23, chromosome 15 open reading frame 23, FLJ14502, HSD11, MGC141728, MGC141729, putative TRAF4-associated factor 1, SKAP, small kinetochore associated protein, TRAF4 associated factor 1, TRAF4AF1 | |
KNSTRN | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title