Learn More
Invitrogen™ SKAP55 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595342
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: PANC whole cell, JURKAT whole cell. IHC: rat spleen tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
Specifications
SKAP55 | |
Polyclonal | |
Unconjugated | |
SKAP1 | |
1700091G21Rik; epididymis secretory sperm binding protein Li 81p; HEL-S-81p; ortholog of human src family associated phosphoprotein 1 SCAP1; pp55; SCAP1; Skap1; Skap55; Skap-55; SKAP55 adapter protein; SKAP55 adaptor protein; src family associated phosphoprotein 1; src family-associated phosphoprotein 1; src kinase associated phosphoprotein 1; src kinase-associated phosphoprotein 1; Src kinase-associated phosphoprotein of 55 kDa | |
Rabbit | |
Affinity chromatography | |
RUO | |
286975, 8631 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q4V7G1, Q86WV1 | |
SKAP1 | |
A synthetic peptide corresponding to a sequence in the middle region of human SKAP55 (163-189aa RMAPHLRRDSKKESCFELTSQDRRSYE). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.