Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117283 Supplier Invitrogen™ Supplier No. PA5117283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTA1
Applications Western Blot
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% glycerol and 0.01% thimerosal; pH 7
Gene ACTA1
Gene Accession No. P68133
Gene Alias AA959943; ACTA; Acta1; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; alpha actin 1; alpha-actin-1; ASMA; beta actin; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; unnamed protein product
Gene Symbols ACTA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human alpha Skeletal Muscle Actin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.