Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SKF 38393 hydrobromide
GSA_VAGREENER_CHOICESDP

Catalog No. 0922100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-221-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-221-00 Supplier R&D Systems™ Supplier No. 0922/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective D1-like agonist

Prototypical D1-like dopamine receptor selective partial agonist (Ki values are 1, ∽ 0.5, ∽ 150, ∽ 5000 and ∽ 1000 nM for D1, D5, D2, D3 and D4 receptors respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 20012-10-6
Molecular Formula C16H18BrNO2

Specifications

Quantity 100 mg
Product Line Tocris
Physical Form Solid
Purity >99%
Molecular Weight (g/mol) 336.229
Chemical Name or Material SKF 38393 hydrobromide
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.