Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SKF 86466 hydrochloride
GREENER_CHOICESDP

Catalog No. 386650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-665-0 50 mg
38-661-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-665-0 Supplier Tocris Bioscience™ Supplier No. 3866/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective α2 antagonist

Potent and selective α2 antagonist at pre- and post-junctional α2-adrenoceptors (Ki values are 13 and 17 nM respectively). Exhibits antihypertensive activity in a rat model.

Chemical Identifiers

CAS 86129-54-6
Molecular Formula C11H15Cl2N

Specifications

Quantity 50 mg
Formula Weight 232.15
Purity >98%
Molecular Weight (g/mol) 232.148
Chemical Name or Material SKF 86466 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.