Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals SKF-96365 hydrochloride, 99+%

Catalog No. AAJ60937MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60937MF 50 mg
AAJ60937MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60937MF Supplier Thermo Scientific Chemicals Supplier No. J60937MF

Inhibits receptor-mediated calcium influx

Inhibits receptor-mediated calcium influx

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130495-35-1
Molecular Formula C22H26N2O3·HCl, C22H27ClN2O3
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Specifications

Quantity 50 mg
Description White
For Use With (Application) Inhibits receptor-mediated calcium influx
Formula Weight 402.92g/mol
Solubility Soluble to 100mM in water
Product Type Signal Transduction Reagent
Form Powder
InChI Key FWLPKVQUECFKSW-UHFFFAOYNA-N
SMILES [H+].[Cl-].COC1=CC=C(CCCOC(CN2C=CN=C2)C2=CC=C(OC)C=C2)C=C1
IUPAC Name hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole chloride
Molecular Weight (g/mol) 402.92
Assay ≥99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.