Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540078
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540078 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540078 Supplier Invitrogen™ Supplier No. PA540078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SKI3 Polyclonal Antibody for Western Blot, ICC/IF

SKI3 is a component of the SKI complex which is composed of SKI2, SKI3 and SKI8. The SKI complex interacts with SKI7, making the link between the SKI complex and the exosome in order to perform mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKI3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4
Gene Ttc37
Gene Accession No. Q6PGP7
Gene Alias KIAA0372; mKIAA0372; Ski3; SKI3 homolog; tetratricopeptide repeat domain 37; tetratricopeptide repeat protein 37; THES; thespin; TPR repeat protein 37; tricho-hepatic-enteric syndrome protein; TTC37
Gene Symbols Ttc37
Host Species Rabbit
Immunogen Recombinant human SKI3 protein.
Purification Method Protein G
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9652
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.