Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558161
Description
Immunogen sequence: QLGLTYWFMG EETRKDKTKA LTHFLKAARL DTYMGKVFCY LGHYYRDVVG DKNRARGCYR KAFELDDTDA ESGAAAVDLS VELEDMEMAL AILTTVTQK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
SKI3 is a component of the SKI complex which is composed of SKI2, SKI3 and SKI8. The SKI complex interacts with SKI7, making the link between the SKI complex and the exosome in order to perform mRNA degradation.
Specifications
| SKI3 | |
| Polyclonal | |
| Unconjugated | |
| Ttc37 | |
| KIAA0372; mKIAA0372; Ski3; SKI3 homolog; tetratricopeptide repeat domain 37; tetratricopeptide repeat protein 37; THES; thespin; TPR repeat protein 37; tricho-hepatic-enteric syndrome protein; TTC37 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9652 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PGP7 | |
| Ttc37 | |
| Recombinant protein corresponding to Human SKI3. Recombinant protein control fragment (Product #RP-96977). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction