Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning Resources Skill Builders Science - Human Body

Click to view available options
Class Size:
1 Student
Description
- In this new take on our Skill Builders! readiness sets, the science is happening inside all of us
- Kids learn about the inner workings of the human body with the help of ageappropriate activities and 3-D pieces for enhanced handson learning.
- Learn the names, functions, and locations of organs, systems, and more
- Includes full-color write-andwipe guide with worksheets.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | STEAM Learning |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction