Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Skin Lotion
SDP

Non-greasy, silicone free moisturizing lotion that helps condition skin

Specifications

Product Type Skin Lotion
Color White
Form Lotion
Quantity 1 Ea.
View More Specs

Products 1
Catalog Number Mfr. No. Volume (English) Packaging Quantity Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Packaging Quantity Volume (Metric) Price Quantity & Availability  
19-053331
SDP
View Documents
GOJO™
814024
5 oz. Tube 1 Ea. 147 mL N/A N/A N/A
Description

Description

  • Contains vitamin E and aloe
800 mL Refill for GOJO™ Bag-in-Box Dispenser
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
Specifications

Specifications

Skin Lotion
White
Lotion
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.