Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596816 Supplier Invitrogen™ Supplier No. PA596816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLPPHISNNY QRLEHFDRIL EMQNCEGRDI PNILDHDLII WFGDMNFRIE DFGLHFVRES IKNRCYGGLW EKDQLSIAKK HDPLLREFQE GRLLFPPTYK FDRNSNDYDT SEKKRKPAWT DRILWRLKRQ PCAGPDTPIP PASHFSLSLR GYSSHMTYGI SDHKPVSGTF DLELKPLVSA PLIVLMPEDL WTVENDMMVS YSSTSDFPSS PWDWIGLYKV GLRDVNDYVS YAWVGDSKVS CSDNLNQVYI DISNIPTTED EFLLCYYSNS LRSVVGISRP FQIPPGSLRE DPLGEAQPQI; Positive Samples: Mouse brain, Rat brain; Cellular Location: Endoplasmic reticulum.

This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKIP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INPP5K
Gene Accession No. Q8C5L6, Q9BT40
Gene Alias C62; Inositol polyphosphate 5-phosphatase K; inositol polyphosphate-5-phosphatase K; Inpp5k; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PI-5-phosphatase related; PPS; putative phosphoinositide 5-phosphatase type II; putative phosphoinositide 5-phosphatase type II; C62; putative PI-5-phosphatase; skeletal muscle and kidney enriched inositol phosphatase; skeletal muscle and kidney enriched inositol polyphosphate phosphatase; skeletal muscle and kidney-enriched inositol phosphatase; SKIP
Gene Symbols INPP5K
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 149-448 of human INPP5K (NP_0576162).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19062, 287533, 51763
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.