Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKIV2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114495
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114495 Supplier Invitrogen™ Supplier No. PA5114495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: Mouse Stomach Tissue Lysate, Human Stomach Tissue Slide.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKIV2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SKIV2L
Gene Accession No. Q15477
Gene Alias 170A; 4930534J06Rik; AW214248; Ddx13; Helicase SKI2W; helicase-like protein; HLP; SKI; SKI2; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; Ski2w; SKIV2; SKIV2L; SKIV2L1; superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like (S. cerevisiae); THES2; W
Gene Symbols SKIV2L
Host Species Rabbit
Immunogen SKI2W antibody was raised against a 16 amino acid peptide near the amino terminus of human SKI2W. The immunogen is located within amino acids 60 - 110 of SKI2W.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108077, 294260, 6499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.