Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKIV2L2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001575JF549
Description
SKIV2L2 Polyclonal antibody specifically detects SKIV2L2 in Human samples. It is validated for Western BlotSpecifications
SKIV2L2 | |
Polyclonal | |
Western Blot | |
ATP-dependent helicase SKIV2L2, Dob1, EC 3.6.1, EC 3.6.4.13, fSAP118, KIAA0052functional spliceosome-associated protein 118, MGC142069, Mtr4superkiller viralicidic activity 2-like 2, superkiller viralicidic activity 2-like 2 (S. cerevisiae) | |
The immunogen recognized by this antibody maps to a region between residues 650 and 700 of human SuperKiller Viralicidic Activity 2-Like 2 using the numbering given in entry NP_056175.2 (GeneID 23517). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23517 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction