Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKIV2L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102808
Description
Antibody detects endogenous levels of total MTR4.
SKIV2L2 may be involved in pre-mRNA splicing.
Specifications
SKIV2L2 | |
Polyclonal | |
Unconjugated | |
MTREX | |
2610528A15Rik; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Dob1; Exosome RNA helicase MTR4; exosome RNA helicase MTR4; superkiller viralicidic activity 2-like 2; fSAP118; functional spliceosome-associated protein 118; KIAA0052; mKIAA0052; Mtr4; Mtr4 exosome RNA helicase; MTREX; RGD1305984; Ski2 like RNA helicase 2; SKIV2L2; Superkiller viralicidic activity 2-like 2; superkiller viralicidic activity 2-like 2 (S. cerevisiae); superkiller viralicidic activity 2-like 2-like; TRAMP-like complex helicase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23517, 365668, 72198 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P42285, Q9CZU3 | |
MTREX | |
A synthesized peptide derived from human SKIV2L2(Accession P42285), corresponding to amino acid residues S758-P808. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction