Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKIV2L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557927
Description
Immunogen sequence: LVVDENGDFR EDNFNTAMQV LRDAGDLAKG DQKGRKGGTK GPSNVFKIVK MIMERNFQPV IIFSFSKKDC EAYALQMTK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
SKIV2L2 may be involved in pre-mRNA splicing.
Specifications
SKIV2L2 | |
Polyclonal | |
Unconjugated | |
MTREX | |
2610528A15Rik; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Dob1; Exosome RNA helicase MTR4; exosome RNA helicase MTR4; superkiller viralicidic activity 2-like 2; fSAP118; functional spliceosome-associated protein 118; KIAA0052; mKIAA0052; Mtr4; Mtr4 exosome RNA helicase; MTREX; RGD1305984; Ski2 like RNA helicase 2; SKIV2L2; Superkiller viralicidic activity 2-like 2; superkiller viralicidic activity 2-like 2 (S. cerevisiae); superkiller viralicidic activity 2-like 2-like; TRAMP-like complex helicase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23517, 365668, 72198 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P42285, Q9CZU3 | |
MTREX | |
Recombinant protein corresponding to Human SKIV2L2. Recombinant protein control fragment (Product #RP-96794). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction