Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKIV2L2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572150
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2150 10 μL
50-157-2149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2150 Supplier Bethyl Laboratories, Inc Supplier No. IHC00470T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SKIV2L2 may be involved in pre-mRNA splicing.

Specifications

Antigen SKIV2L2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SKIV2L2
Gene Accession No. P42285, Q9CZU3
Gene Alias ATP-dependent helicase SKIV2L2, ATP-dependent RNA helicase DOB1, ATP-dependent RNA helicase SKIV2L2, Dob1, fSAP118, functional spliceosome-associated protein 118, KIAA0052, Mtr4, superkiller viralicidic activity 2-like 2 (S. cerevisiae), TRAMP-like complex helicase
Gene Symbols SKIV2L2
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23517, 72198
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.