Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKOR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139977
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GPDGEQPTGP PSATSSGADG PANSPDGGSP RPRRRLGPPP AGRPAFGDLA AEDLVRRPER SPPSGGGGYE LR.
SKOR1 acts as a transcriptional corepressor of LBX1. Inhibits BMP signaling.
Specifications
SKOR1 | |
Polyclonal | |
Unconjugated | |
SKOR1 | |
AV273001; C230094B15Rik; corepressor for LBX1; CORL1; functional smad suppressing element 15; functional Smad-suppressing element on chromosome 15; FUSSEL15; fussel-15; fussel-15 homolog; ladybird homeobox 1 homolog corepressor 1; ladybird homeobox corepressor 1; ladybird homeobox corepressor 1; LOW QUALITY PROTEIN: SKI family transcriptional corepressor 1; LBX1 corepressor 1; Lbxcor1; Lbxcor1 homolog; RGD1307687; SKI family transcriptional corepressor 1; Skor1; Transcriptional corepressor Corl1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
390598 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P84550 | |
SKOR1 | |
Recombinant protein corresponding to Human SKOR1. Recombinant protein control fragment (Product #RP-109249). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction