Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKOR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561186 Supplier Invitrogen™ Supplier No. PA561186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASSPLPGPN DILLASPSSA FQPDTLSQPR PGHANLKPNQ VGQVILYGIP IVS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Exhibits transcriptional repressor activity. Acts as a TGF-beta antagonist in the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKOR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SKOR2
Gene Accession No. A7M7C7
Gene Alias CH18515; Corl2; EG664805; functional smad suppressing element 18; functional Smad suppressor element on chromosome 18; functional Smad-suppressing element on chromosome 18; FUSSEL18; Fussel-18; fussel-18 homolog; Gm7348; ladybird homeobox corepressor 1-like protein; LBX1 corepressor 1-like protein; SKI family transcriptional corepressor 2; Skor2; Transcriptional corepressor Corl2
Gene Symbols SKOR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SKOR2. Recombinant protein control fragment (Product #RP-98298).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 664805
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.