Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKP1 Polyclonal Antibody

Catalog No. PIPA5121607
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121607 20 μL
PIPA5119432 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121607 Supplier Invitrogen™ Supplier No. PA5121607
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SKP1 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a component of SCF complexes, which are composed of this protein, cullin 1, a ring-box protein, and one member of the F-box family of proteins. This protein binds directly to the F-box motif found in F-box proteins. SCF complexes are involved in the regulated ubiquitination of specific protein substrates, which targets them for degradation by the proteasome. Specific F-box proteins recognize different target protein(s), and many specific SCF substrates have been identified including regulators of cell cycle progression and development. Studies have also characterized the protein as an RNA polymerase II elongation factor. Alternative splicing of this gene results in two transcript variants. A related pseudogene has been identified on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SKP1
Gene Accession No. P63208, Q6PEC4
Gene Alias 15kDa; 2610043E24Rik; 2610206H23Rik; cyclin A/CDK2-associated p19; cyclin A/CDK2-associated protein p19; cyclin-A/CDK2-associated protein p19; EMC19; MGC34403; OCP2; OCP-2; OCP-II; OCP-II protein; organ of Corti protein 2; Organ of Corti protein II; p19A; p19Skp1; polypeptide 1-like; RNA polymerase II elongation factor-like protein; RNA polymerase II elongation factor-like protein OCP2; SIII; Skp1; Skp1a; S-phase kinase-associated protein 1; S-phase kinase-associated protein 1A; TCEB1L; transcription elongation factor B polypeptide 1-like
Gene Symbols SKP1
Host Species Rabbit
Immunogen Recombinant protein S-Phase Kinase Associated Protein 1. The antigen corresponds to amino acid range 1-163 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110259483, 287280, 6500
Target Species Human, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.