Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKP1 Recombinant Rabbit Monoclonal Antibody (JM27-37)
GREENER_CHOICE

Catalog No. PIMA532744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532744 Supplier Invitrogen™ Supplier No. MA532744
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a component of SCF complexes, which are composed of this protein, cullin 1, a ring-box protein, and one member of the F-box family of proteins. This protein binds directly to the F-box motif found in F-box proteins. SCF complexes are involved in the regulated ubiquitination of specific protein substrates, which targets them for degradation by the proteasome. Specific F-box proteins recognize different target protein(s), and many specific SCF substrates have been identified including regulators of cell cycle progression and development. Studies have also characterized the protein as an RNA polymerase II elongation factor. Alternative splicing of this gene results in two transcript variants. A related pseudogene has been identified on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JM27-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene SKP1
Gene Accession No. P63208, Q6PEC4, Q9WTX5
Gene Alias 15kDa; 2610043E24Rik; 2610206H23Rik; cyclin A/CDK2-associated p19; cyclin A/CDK2-associated protein p19; cyclin-A/CDK2-associated protein p19; EMC19; MGC34403; OCP2; OCP-2; OCP-II; OCP-II protein; organ of Corti protein 2; Organ of Corti protein II; p19A; p19Skp1; polypeptide 1-like; RNA polymerase II elongation factor-like protein; RNA polymerase II elongation factor-like protein OCP2; SIII; Skp1; Skp1a; S-phase kinase-associated protein 1; S-phase kinase-associated protein 1A; TCEB1L; transcription elongation factor B polypeptide 1-like
Gene Symbols SKP1, Skp1a
Host Species Rabbit
Immunogen Synthetic peptide within Human Skp1 aa 106-155.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21402, 287280, 6500
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.