Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Skp2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297674MFV610
Description
Skp2 Polyclonal antibody specifically detects Skp2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Skp2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
CDK2/cyclin A-associated protein p45, cyclin A/CDK2-associated protein p45, Cyclin-A/CDK2-associated protein p45, FBL1, F-box protein Skp2, F-box/LRR-repeat protein 1, FBXL1MGC1366, FLB1, p45skp2, S-phase kinase-associated protein 2, S-phase kinase-associated protein 2 (p45) | |
Produced in rabbits immunized with E. coli-derived Human Skp2 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
6502 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction