Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKP2 Monoclonal Antibody (6G9D10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515470
Description
MA5-15470 targets SKP2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15470 immunogen is purified recombinant fragment of SKP2 (aa1-130) expressed in E. Coli.
Skp2 (S-phase Kinase-associated Protein 2) belongs to the family of F-box proteins that interact with the Cyclin A-Cdk2 complex. Skp2 is essential for the G1-S transition in both transformed cells and diploid fibroblasts. Biochemical and genetic experiments have demonstrated that Skp2 is required for the ubiquitination and consequent degradation of p27 in cultured mammalian cells and in vitro reconstitution assays. In normal tissues, Skp2 is expressed in tonsil and placenta. In tumor tissues, Skp2 is expressed widely, in some colon, prostate, pancreas, and skin cancers, with high expression in lung, breast, ovarian, and endometrial cancers. Research suggests that Skp2 plays a role in oncogenesis, particularly in the pathogenesis of lymphomas and breast cancers.
Specifications
| SKP2 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q13309 | |
| SKP2 | |
| Purified recombinant fragment of SKP2 (aa1-130) expressed in E. Coli. | |
| RUO | |
| 6502 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 6G9D10 | |
| Unconjugated | |
| SKP2 | |
| 4930500A04Rik; CDK2/cyclin A-associated protein p45; cyclin A/CDK2-associated protein p45; Cyclin-A/CDK2-associated protein p45; FBL1; F-box protein Skp2; F-box/LRR-repeat protein 1; F-box/WD-40 protein 1; FBXL1; FLB1; FWD1; MGC1366; OTTHUMP00000221389; p45; p45skp2; RGD1562456; Skp2; S-phase kinase-associated protein 2; S-phase kinase-associated protein 2 (p45); S-phase kinase-associated protein 2, E3 ubiquitin protein ligase | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction