Learn More
MSA™ Skullgard™ Protective Caps and Hats
Catalog No. 189993463 Shop All MSA Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Natural Tan
Orange
Red
Red-Orange
White
Yellow
Yellow-Orange
Head Style:
Front Brim
Full Brim
Suspension Type:
Fas-Trac Ratchet
Fas-Trac Ratchet with attached Welders’ Lugs
Staz-On Suspension
Staz-On Suspension with attached lamp bracket and cord holder
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Head Style Suspension Type
18-999-3463 White Full Brim Staz-On Suspension
19-043746 Yellow Full Brim Staz-On Suspension
18-999-3474 White Front Brim Fas-Trac Ratchet
18-999-3494 Natural Tan Front Brim Fas-Trac Ratchet
18-999-3441 Natural Tan Full Brim Staz-On Suspension
18-999-3506 Red Front Brim Staz-On Suspension
18-999-3508 Blue Front Brim Staz-On Suspension
18-999-3475 Blue Front Brim Fas-Trac Ratchet
19-088-1044 Green Full Brim Staz-On Suspension
19-043665 Yellow-Orange Front Brim Staz-On Suspension
19-815-467 Blue Full Brim Staz-On Suspension
18-999-3451 Natural Tan Full Brim Fas-Trac Ratchet
19-064431 Green Front Brim Staz-On Suspension
18-999-3504 Orange Front Brim Staz-On Suspension
18-999-3447 White Full Brim Fas-Trac Ratchet
19-815-468 Orange Full Brim Staz-On Suspension
18-999-3452 Natural Tan Front Brim Staz-On Suspension with attached lamp bracket and cord holder
19-043069 White Front Brim Staz-On Suspension
18-999-3507 Gray Front Brim Staz-On Suspension
18-999-3461 Red-Orange Front Brim Staz-On Suspension
18-999-3449 Natural Tan Front Brim Fas-Trac Ratchet with attached Welders’ Lugs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Do not mix suspension and helmet sizes. Make sure the sizes match (example: standard size suspension with standard size helmet). Use only MSA suspensions on an MSA helmet. Failure to comply with above will reduce the energy-absorbing ability of the protective helmet, which can result in serious injury or death.
Catalog No. 18-999-3463 Supplier MSA™ Supplier No. 454665
Add to Cart
Add to Cart
Do not mix suspension and helmet sizes. Make sure the sizes match (example: standard size suspension with standard size helmet). Use only MSA suspensions on an MSA helmet. Failure to comply with above will reduce the energy-absorbing ability of the protective helmet, which can result in serious injury or death.
Suitable for use in steel mills
Heavy-duty construction makes cap and hat preferred for use in steel mills and other environments that combine elevated temperatures and risk of injury from falling objects.
  • Shells are hand-made in the USA of layered, resin-impregnated textiles laminated under pressure
  • Wishstands radiant heat loads up to, but not exceeding, 350°F
  • Color: Natural tan
Compliance
Third-party certified by SEI to meet the Type I, Class G requirements of ANSI/ISEA Z89.1.
Specifications
Certifications/Compliance Meets ANSI/ISEA Z89.1-2009; Class G.
Color White
Size Standard (6.5 to 8 in.)
Suspension Type Staz-On Suspension
Head Style Full Brim
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.