Learn More
Pyramex™ SL Series Cap Style Hard Hat
Exceeds ANSI Z89.1, Type 1, Class C, G, and E for industrial head protection; CSA Z94.1-15
$8.85 - $244.83
Specifications
Material HDPE
View More Specs
Products 28
Catalog Number Mfr. No. Product Type Color Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Color Price Quantity & Availability  
19-155-372
View Documents
Pyramex™
HP14020
4-Point Snap Lock Red
Case of 16 for $121.19
Only null left
Add to Cart
19-155-373
View Documents
Pyramex™
HP14030
4-Point Snap Lock Yellow
Case of 16 for $151.46
Only null left
Add to Cart
19-155-374
View Documents
Pyramex™
HP14031
4-Point Snap Lock Hi-vis Lime
Case of 16 for $189.42
Only null left
Add to Cart
19-155-375
View Documents
Pyramex™
HP14035
4-Point Snap Lock Green
Case of 16 for $151.46
Only null left
Add to Cart
19-155-376
View Documents
Pyramex™
HP14040
4-Point Snap Lock Orange
Case of 16 for $121.19
Only null left
Add to Cart
19-155-377
View Documents
Pyramex™
HP14060
4-Point Snap Lock Blue
Case of 16 for $151.46
Only null left
Add to Cart
19-155-378
View Documents
Pyramex™
HP14062
4-Point Snap Lock Light Blue
Each for $8.85
Pack of 16 EA for $119.10
Only null left
Add to Cart
19-155-381
View Documents
Pyramex™
HP14111
4-Point Ratchet Black
Case of 16 for $185.40
Only null left
Add to Cart
19-155-385
View Documents
Pyramex™
HP14131
4-Point Ratchet Hi-vis Lime
Case of 16 for $231.47
Only null left
Add to Cart
19-155-387
View Documents
Pyramex™
HP14140
4-Point Ratchet Orange
Case of 16 for $185.40
Only null left
Add to Cart
19-155-388
View Documents
Pyramex™
HP14141
4-Point Ratchet Hi-vis Orange
Case of 16 for $231.47
Only null left
Add to Cart
19-155-390
View Documents
Pyramex™
HP14162
4-Point Ratchet Light Blue
Case of 16 for $185.40
Only null left
Add to Cart
19-155-391
View Documents
Pyramex™
HP14165
4-Point Ratchet Dark Blue
Case of 16 for $185.40
Only null left
Add to Cart
19-155-392
View Documents
Pyramex™
HP14170
4-Point Ratchet Hi-vis Pink
Case of 16 for $238.07
Only null left
Add to Cart
19-155-393
View Documents
Pyramex™
HP16010
6-Point Snap Lock White
Case of 16 for $165.94
Only null left
Add to Cart
19-155-394
View Documents
Pyramex™
HP16020
6-Point Snap Lock Red
Case of 16 for $162.00
Only null left
Add to Cart
19-155-395
View Documents
Pyramex™
HP16030
6-Point Snap Lock Yellow
Case of 16 for $162.00
Only null left
Add to Cart
19-155-397
View Documents
Pyramex™
HP16040
6-Point Snap Lock Orange
Case of 16 for $162.00
Only null left
Add to Cart
19-155-400
View Documents
Pyramex™
HP16110
6-Point Ratchet White
Case of 16 for $199.96
Only null left
Add to Cart
19-155-401
View Documents
Pyramex™
HP16120
6-Point Ratchet Red
Case of 16 for $199.96
Only null left
Add to Cart
19-155-402
View Documents
Pyramex™
HP16130
6-Point Ratchet Yellow
Case of 16 for $205.17
Only null left
Add to Cart
19-155-403
View Documents
Pyramex™
HP16135
6-Point Ratchet Green
Case of 16 for $199.96
Only null left
Add to Cart
19-155-404
View Documents
Pyramex™
HP16140
6-Point Ratchet Orange
Case of 16 for $199.96
Only null left
Add to Cart
19-155-405
View Documents
Pyramex™
HP16141
6-Point Ratchet Hi-vis Orange
Case of 16 for $244.83
Only null left
Add to Cart
19-155-406
View Documents
Pyramex™
HP16160
6-Point Ratchet Blue
Case of 16 for $199.96
Only null left
Add to Cart
19-151-528
View Documents
Pyramex™
HP14010
4-Point Snap Lock White
Case of 16 for $155.40
Only null left
Add to Cart
19-155-396
View Documents
Pyramex™
HP16035
6-Point Snap Lock Green N/A N/A N/A
19-155-398
View Documents
Pyramex™
HP16060
6-Point Snap Lock Blue N/A N/A N/A
Description
Description
  • Low profile design
  • Tested and marked for reverse donning
  • Shell constructed from High Density Polyethylene materials
  • Rain trough on sides and back of helmet channel moisture away
  • Soft brow pad is replaceable. Replaceable suspensions and headbands also available
  • Snap Lock suspension features 'tuck away' adjustments for all day, hassle free wear
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
  • Universal accessory slots fit cap mounted ear muffs and accessories
  • Hats also accommodate chin straps
  • Testing: Impact, Penetration, Flammability, Electrical Insulation, Marking and Humidity
  • Specifications
    Specifications
    HDPE
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.