Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLA/LP Antibody, ProSci
SDP

Catalog No. 16831737
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-831-737 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-831-737 Supplier ProSci LLC Supplier No. 295150100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLA/LP Polyclonal Antibody specifically detects SLA/LP in Human, Mouse, Rat, Dog samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Specifications

Antigen SLA/LP
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution SLA/LP antibody can be used for detection of SLA/LP by ELISA at 1:312500. SLA/LP antibody can be used for detection of SLA/LP by western blot at 1.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_722547
Gene Alias SLA/LP, LP, SLA, PCH2D, SLA/LP
Host Species Rabbit
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human SLA/LP.
Purification Method Antibody is purified by peptide affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51091
Target Species Human, Mouse, Rat, Dog
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store SLA/LP antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Other
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.