Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542141
Description
Peptide sequence: PEGTENPLGV DESLFSYGLR ESIASYLSLT SEDNTSFDRK KKSISLMYGG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SLAP also known as Src-like-adapter protein 1 is a 276 amino acid containing adapter protein, functioning as a negative regulator of T-cell receptor (TCR) signaling. SLAP belonging to the SRC family of cytoplasmic tyrosine kinases contains a SH2 domain and a SH3 domain. This cytoplasmic protein colocalizes with endosomes and is known to inhibit T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. It acts as a negative regulator of positive selection and mitosis of T-cells. SLAP may act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins. Expression of SLAP is weak in heart, adult brain, placenta, liver, skeletal muscle, kidney and pancreas but is high in lung and fetal brain.
Specifications
| SLA | |
| Polyclonal | |
| Unconjugated | |
| SLA | |
| hSLAP; Sla; SLA1; SLAP; Slap1; Slap-1; Src like adaptor; src-like adaptor; src-like-adapter; Src-like-adapter protein 1; Src-like-adaptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6503 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13239 | |
| SLA | |
| Synthetic peptide directed towards the middle region of human SLA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction