Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLA2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SLA2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179830
SDP
View Documents
Novus Biologicals
NBP179830
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SLA2 Polyclonal specifically detects SLA2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLA2
Polyclonal
Rabbit
NP_115590
84174
Synthetic peptide directed towards the middle region of human SLA2The immunogen for this antibody is SLA2. Peptide sequence WLYEGLSREKAEELLLLPGNPGGAFLIRESQTRRGSYSLSVRLSRPASWD.
Primary
Western Blot
Unconjugated
RUO
C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2
SLA2
IgG
28 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.