Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLA2 Antibody (OTI3A7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274210AF532
Description
SLA2 Monoclonal specifically detects SLA2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
SLA2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SLA2 | |
Full length recombinant protein of human SLA2 (NP_115590) produced in HEK293T cells. | |
Immunology, Signal Transduction | |
84174 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI3A7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2 | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction