Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLA2 Antibody (OTI3A7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274210PCP
Description
SLA2 Monoclonal specifically detects SLA2 in Human samples. It is validated for Flow Cytometry.Specifications
| SLA2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| SLA2 | |
| Full length recombinant protein of human SLA2 (NP_115590) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI3A7 | |
| Flow Cytometry | |
| C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2 | |
| Mouse | |
| Affinity Purified | |
| Immunology, Signal Transduction | |
| 84174.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction