Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLA2 Antibody (Z30P1F12*F4), Janelia Fluor™ 646, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP250293JF646

Catalog No. NBP50293646


Only null left
Add to Cart

Description

Description

SLA2 Monoclonal specifically detects SLA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

SLA2
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
SLA2
Peptide Sequence - LRESLSFY
0.1 mL
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
Z30P1F12*F4
Western Blot, ELISA, Immunohistochemistry
C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2
Mouse
Protein A purified
Immunology, Signal Transduction
84174
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.