Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLA2 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505143S


Description
SLA2 Monoclonal antibody specifically detects SLA2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
SLA2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H6Q3 | |
Mouse | |
Affinity chromatography | |
RUO | |
84174 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G3 | |
Unconjugated | |
SLA2 | |
A930009E21Rik; AI430952; C20orf156; MARS; Modulator of antigen receptor signaling; modulator of antigen receptor signaling MARS; RGD1562071; Sla2; SLAP2; SLAP-2; Src like adaptor 2; src-like adapter protein 2; Src-like adapter protein-2; src-like-adapter 2; Src-like-adaptor 2 | |
Full length human recombinant protein of human SLA2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction