Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLA2 Monoclonal Antibody (OTI3A7)
GREENER_CHOICE

Catalog No. PIMA526059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526059 Supplier Invitrogen™ Supplier No. MA526059
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLA2 Monoclonal Antibody for Western Blot, IHC (P), Flow

Adapter protein, which negatively regulates T-cell receptor (TCR) signaling. Inhibits T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. May act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLA2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLA2
Gene Accession No. Q9H6Q3
Gene Alias A930009E21Rik; AI430952; C20orf156; MARS; Modulator of antigen receptor signaling; modulator of antigen receptor signaling MARS; RGD1562071; Sla2; SLAP2; SLAP-2; Src like adaptor 2; src-like adapter protein 2; Src-like adapter protein-2; src-like-adapter 2; Src-like-adaptor 2
Gene Symbols SLA2
Host Species Mouse
Immunogen Full length human recombinant protein of SLA2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84174
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.