Learn More
Invitrogen™ SLA2 Polyclonal Antibody

Description
Immunogen sequence: HYSELADDICC LLKEPCVLQR AGPLPGKDIP LPVTVQRTPL NWKELDSSLL F Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.
Specifications
Specifications
| Antigen | SLA2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLA2 |
| Gene Accession No. | Q9H6Q3 |
| Gene Alias | A930009E21Rik; AI430952; C20orf156; MARS; Modulator of antigen receptor signaling; modulator of antigen receptor signaling MARS; RGD1562071; Sla2; SLAP2; SLAP-2; Src like adaptor 2; src-like adapter protein 2; Src-like adapter protein-2; src-like-adapter 2; Src-like-adaptor 2 |
| Gene Symbols | SLA2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.