Learn More
Invitrogen™ SLA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588777
Description
Immunogen sequence: MGSLPSRRKS LPSPSLSSSV QGQGPVTMEA ERSKATAVAL GSFPAGGPAE LSLRLGEPLT IVSEDGDWWT VLSEVSGREY NIPSVHVAKV SHGWLYEGLS REKAEELLLL PGNPGGAFLI RESQTRRGSY SLSVRLSRPA SWDRIRHYRI HCLDNGWLYI; Positive Samples: K-562, A-431, HT-29; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic vesicle.
Adapter protein, which negatively regulates T-cell receptor (TCR) signaling. Inhibits T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. May act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins.
Specifications
SLA2 | |
Polyclonal | |
Unconjugated | |
SLA2 | |
A930009E21Rik; AI430952; C20orf156; MARS; Modulator of antigen receptor signaling; modulator of antigen receptor signaling MARS; RGD1562071; Sla2; SLAP2; SLAP-2; Src like adaptor 2; src-like adapter protein 2; Src-like adapter protein-2; src-like-adapter 2; Src-like-adaptor 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84174 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.01 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9H6Q3 | |
SLA2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human SLA2 (NP_1155901). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.