Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAIN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204062
Description
SLAIN1 Polyclonal specifically detects SLAIN1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SLAIN1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
SLAIN motif family, member 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
122060 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
NP_001035243.1 | |
SLAIN1 | |
The immunogen recognized by this antibody maps to a region between residue 518 and 568 of human SLAIN Motif Family, Member 1 using the numbering given in entry NP_001035243.1 (GeneID 122060). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction