Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLAIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598761
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598761 Supplier Invitrogen™ Supplier No. PA598761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLAIN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SLAIN1 (SLAIN Motif Family Member 1) is a Protein Coding gene. An important paralog of this gene is SLAIN2. Microtubule plus-end tracking protein that might be involved in the regulation of cytoplasmic microtubule dynamics, microtubule organization and microtubule elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLAIN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLAIN1
Gene Accession No. Q8ND83
Gene Alias 9630044O09Rik; AA675320; AW742596; C13orf32; RGD1308626; SLAIN motif family member 1; SLAIN motif family, member 1; SLAIN motif-containing protein 1; SLAIN1
Gene Symbols SLAIN1
Host Species Rabbit
Immunogen Recombinant Human SLAIN motif-containing protein 1 protein (43-136AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122060
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.